Would you like to log in again?
6×3 X-Gen Goggles
Supplier: Univet
MDU6004SC MDU6003SC
77393-768EA 11 USD InStock
77393-768 77393-766
6×3 X-Gen Goggles
Goggles
6×3 X-Gen goggles feature a panoramic lens that easily integrates with prescription glasses.
  • Lens treatment: anti-fog and anti-scratch (double coating)
  • Interchangeable lens system
  • Frame engineered for soft facial support
  • Compatible with filtering half masks
6×3 X-Gen goggles feature a lens with anti-fog and anti-scratch ultra performing Vanguard UDC treatment. They are certified according to EN166 standard, level 3 and 4 (EU) for protection against droplets, splashes, and large dust particles.
Ordering information: The 6×3 goggle face shield is available separately as a face protection accessory for this goggle series.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty