Would you like to log in again?
6×1 X-Gen Goggle
Supplier: Univet
MDU6005S
77393-770PK 130 USD InStock
77393-770
6×1 X-Gen Goggle
Goggles
6×1 X-Gen goggles feature a broad lens for a wide field of view with zero distortions.
  • Lens treatment: anti-fog and anti-scratch (double coating), plus UV380
  • Gunmetal/green frame color
  • Clear lens
Goggles are designed with an internal soft rubber to ensure a comfortable fit with no inconvenient high pressure points. They feature an interchangeable system between temples and elastic band (included in the package). The temples have rubber terminals and are adjustable in length and inclination. The lens is treated with VPlus anti-fog and anti-scratch enhancement.
Certifications: Certified according to EN 166 standard, level 3 and 4 (EU) for protection against droplets, splashes, and large dust particles.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty