Would you like to log in again?
Wax Dispenser
Supplier: Four E's USA
WXD100101
77394-612EA 1168.97 USD InStock
77394-612
Wax Dispenser
Paraffin Wax Trimmers and Dispensers
The paraffin wax dispenser features a fast heat-up time and a heated spout to provide a seamless dispensing experience.
  • Fast and uniform wax heating
  • Digital display shows temperature and timer
  • Adjustable spout heating intensity with rear knob
  • Alarm light activates when temperature exceeds set value by 5°
  • Settings are saved and automatically restored upon restart
The 10 L tank provides ample capacity while saving benchtop space. The antimicrobial 304 stainless steel inner chamber is fully insulated for safe handling. Inside the inner chamber there is a filter screen to prevent particles from entering and clogging the spout, and the non-drip spout is heated to prevent wax buildup. The faucet has 3 functional positions: off, on-demand dispense, and continuous flow.
Delivery information: Typically in stock, ready to ship.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty