Would you like to log in again?
Seamless Assembly Master Mix (2X)
Supplier: ReVacc Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Q72R56-250UL Q72R56-1ML
77394-702EA 1090 USD InStock
77394-702 77394-704
Seamless Assembly Master Mix (2X)
Nucleic Acid Reagents Cloning Kits and Ligation
This Seamless Assembly Master Mix (2X) uses Gibson cloning principle to allow efficiently assembly of multiple DNA fragments, regardless of fragment length or end compatibility. The insert and the linearized vector need the overlapped sequences of 15 bp - 20 bp on both 5' - and 3' -end, respectively. A positive control with the mixture of insert and linearized vector is included.
  • Assembly is a one-step mixture with the incubation up to 1 hour at 50 °C
  • No restriction enzyme digestion, gel extraction or overnight ligation
  • PCR clean-up step is optional, even clean-up will increase cloning efficiency
  • Adjacent DNA fragment needs shared overlaps
  • High transformation efficiencies for inserts up to 20 kb
  • Easily manipulation of up to 4 DNA fragments into linearized vector
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty