Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
BioX Tissue Culture Flasks
Supplier: TradeWinds (TWD)
BioX
BX07-8025 BX07-8075 BX07-8300 BX07-8182
77394-954CS 205.62 USD InStock
77394-954 77394-956 77394-958 77394-960
BioX Tissue Culture Flasks
Flasks Culture Flasks
Ideal for the cultivation and observation of sensitive cells.
  • Tilted neck
  • PTFE hydrophobic vent cap
  • Marked volume scales on both sides
  • Frosted writing area
  • DNase/RNase, non-pyrogenic
  • Highly transparent polystyrene
The angled neck allows the user to access the entire surface area. Easy sample identification due to the molded graduation and frosted writing patch.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty