Would you like to log in again?
BioX Vacuum Filtration Systems, Sterile
Supplier: TradeWinds (TWD)
BioX
BX15-4500 BX15-4250 BX15-4150 BX15-4000
77394-964CS 106.9 USD InStock
77394-964 77394-966 77394-968 77394-962
BioX Vacuum Filtration Systems, Sterile
Filters Bottle Top Filters
Systems for vacuum filtration of solutions including buffers, cell culture media, or other fluids.
  • PES membrane
  • 0.22 µm pore size
  • Lightweight heavy wall construction
  • Engraved graduations
  • Non-pyrogenic
Vacuum filters are very useful for the separation and purification of large volumes of samples.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty