Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Super-Duty 88 GSM SMS Autoclavable Bowl Covers with Steam Indicator

Supplier: Keystone Cleanroom Products

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

BWL-KCRP-600-60 BWL-KCRP-600-30 BWL-KCRP-600-20 BWL-KCRP-600-24 BWL-KCRP-600-54 BWL-KCRP-600-48 BWL-KCRP-600-36 BWL-KCRP-600-42 BWL-KCRP-600-50 BWL-KCRP-600-40
77395-104CS 1302 USD InStock
77395-104 77395-102 77395-110 77395-100 77395-098 77395-096 77395-094 77395-092 77395-108 77395-106
Super-Duty 88 GSM SMS Autoclavable Bowl Covers with Steam Indicator
Covers
This 88 g/m² SMS autoclavable bowl cover provides a microbial barrier that is ideal for pharmaceutical production applications.

  • Hydrophobic, tear-resistant, and puncture-resistant
  • Creates secondary level of protection during aseptic presentation and setup of equipment
  • Designed for autoclave cycles (121 to 135 °C)
  • Manufactured and packed in a certified ISO 8 cleanroom
  • Made in USA

Eliminate the need for cutting sheets of sterilization wrapping materials and securing them with autoclave tape and rubber bands. After the outer bag is removed, this stopper bowl cover should remain in place, providing protection from microbial and particulate contamination while the remaining set-up activities are performed. Used in conjunction with a secondary autoclave bag, it creates an effective wrapping system.

Certifications: Certificates of conformance are sent with every shipment.

Packaging: Cleanroom packed in sealed, labeled poly-bags with the lot number and date of manufacture. Covers are packed 1 per bag.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty