Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Blue Heavy-Duty Autoclavable SMS Muff Covers

Supplier: Keystone Cleanroom Products

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

MFKCRP300-06 MFKCRP300-09 MFKCRP300-03 MFKCRP300-08 MFKCRP300-12 MFKCRP300-04 MFKCRP300-18 MFKCRP300-05 MFKCRP300-07 MFKCRP300-16
77395-114CS 1364 USD InStock
77395-114 77395-126 77395-112 77395-124 77395-122 77395-130 77395-120 77395-118 77395-116 77395-128
Blue Heavy-Duty Autoclavable SMS Muff Covers
Covers
This 50 g/m² SMS autoclavable muff cover provides an excellent microbial barrier that is ideal for pharmaceutical production applications.

  • Eliminates autoclave tape residue
  • Hydrophobic, tear-resistant, and puncture-resistant
  • Creates secondary level of protection during aseptic presentation and setup of equipment
  • Designed for autoclave cycles (121 to 135 °C)
  • Manufactured and packed in a certified ISO 8 cleanroom
  • Used in conjunction with a secondary autoclave bag, it creates an effective wrapping system
  • Made in USA

Eliminate the need for cutting sheets of sterilization wrapping materials and securing them with autoclave tape and rubber bands. Cost-effective, time-saving approach standardizes the equipment preparation process, minimizes particulate generation, while also ensuring good aseptic technique upon removal after sterilization.

Certifications: Certificates of conformance are sent with every shipment.

Packaging: Cleanroom packed in sealed, labeled poly-bags with the lot number and date of manufacture. Covers are packed 100 per bag.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty