Would you like to log in again?
Blue Super-Duty Autoclavable SMS Muff Covers
Supplier: Keystone Cleanroom Products
MFKCRP600-03 MFKCRP600-06 MFKCRP600-16 MFKCRP600-04 MFKCRP600-18 MFKCRP600-09 MFKCRP600-12 MFKCRP600-05 MFKCRP600-08 MFKCRP600-07
77395-136CS 1627.5 USD InStock
77395-136 77395-148 77395-134 77395-146 77395-132 77395-144 77395-142 77395-150 77395-140 77395-138
Blue Super-Duty Autoclavable SMS Muff Covers
Covers
This 88 g/m² SMS autoclavable muff cover provides an excellent microbial barrier that is ideal for pharmaceutical production applications.
  • Eliminates autoclave tape residue
  • Hydrophobic, tear-resistant, and puncture-resistant
  • Creates secondary level of protection during aseptic presentation and setup of equipment
  • Designed for autoclave cycles (121 to 135 °C)
  • Manufactured and packed in a certified ISO 8 cleanroom
  • Used in conjunction with a secondary autoclave bag, it creates an effective wrapping system
  • Made in USA
Eliminate the need for cutting sheets of sterilization wrapping materials and securing them with autoclave tape and rubber bands. Cost-effective, time-saving approach standardizes the equipment preparation process, minimizes particulate generation, while also ensuring good aseptic technique upon removal after sterilization.
Certifications: Certificates of conformance are sent with every shipment.
Packaging: Cleanroom packed in sealed, labeled poly-bags with the lot number and date of manufacture. Covers are packed 100 per bag.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty