Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Blue Super-Duty Autoclavable SMS Muff Covers

Supplier: Keystone Cleanroom Products

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

MFKCRP600-03 MFKCRP600-06 MFKCRP600-16 MFKCRP600-04 MFKCRP600-18 MFKCRP600-09 MFKCRP600-12 MFKCRP600-05 MFKCRP600-08 MFKCRP600-07
77395-136CS 1627.5 USD InStock
77395-136 77395-148 77395-134 77395-146 77395-132 77395-144 77395-142 77395-150 77395-140 77395-138
Blue Super-Duty Autoclavable SMS Muff Covers
Covers
This 88 g/m² SMS autoclavable muff cover provides an excellent microbial barrier that is ideal for pharmaceutical production applications.

  • Eliminates autoclave tape residue
  • Hydrophobic, tear-resistant, and puncture-resistant
  • Creates secondary level of protection during aseptic presentation and setup of equipment
  • Designed for autoclave cycles (121 to 135 °C)
  • Manufactured and packed in a certified ISO 8 cleanroom
  • Used in conjunction with a secondary autoclave bag, it creates an effective wrapping system
  • Made in USA

Eliminate the need for cutting sheets of sterilization wrapping materials and securing them with autoclave tape and rubber bands. Cost-effective, time-saving approach standardizes the equipment preparation process, minimizes particulate generation, while also ensuring good aseptic technique upon removal after sterilization.

Certifications: Certificates of conformance are sent with every shipment.

Packaging: Cleanroom packed in sealed, labeled poly-bags with the lot number and date of manufacture. Covers are packed 100 per bag.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty