Would you like to log in again?
VWR® Wipa-Boots, Sterile Sampling Boot Covers
Supplier: Avantor
VWR®
KSS-67410-VW1 KSS-67410-VW1
77395-160CS 139.61 USD InStock
CA77395-160 77395-160
VWR® Wipa-Boots, Sterile Sampling Boot Covers
Workwear
A kit consisting of sterile woven cotton slippers inserted in a sterile bag. The kit allows for large area sampling at ground level.
  • Ideal for sample collection and inspection of surfaces in contact with livestock
  • Easy to use and highly versatile
  • Simultaneous sample collection and inspection
  • Sterile and biocide-free
  • Fits any work boots size
Wipa-boots are impregnated with a solution that allows transportation of different pathogens. A major concern in the livestock industry is the prevention, detection, and control of infections by pathogens like viruses (H1N1, African swine fever, PRRS, Circovirus, Newcastle) and bacteria (Salmonella and E. coli including STEC). This product is a quick and simple solution to establish an innocuous environment in livestock housing, animal transportation vehicles, and their washing stations.
Acting as a quick and simple sterile sampling method, Wipa-boots allow sample collection to occur simultaneously during routine walking inspections. The kit includes a sterile bag and a pair of boot covers, pre-moistened in 23 ml of tryptone salt. This solution allows the transportation of different pathogens, for reliable and safe analysis.
Certifications: Sterility is provided by irradiation. The sterilization dose ensuring a sterility assurance level (SAL) of 10⁻³ has been established according to ISO 11137.
Caution: Upon receipt, store between 2 and 8 °C.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty