Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aqua Block™ Tris

Supplier: Arlington Scientific
Aquatic Block™
PP82T-500 PP82T-125
77395-188EA 110 USD InStock
77395-188 77395-186
Aqua Block™ Tris
Buffers Electrophoresis Buffers Blocking Buffers
A non-mammalian blocking reagent supplied in tris for diagnostic systems involving membrane or other solid phase material.

  • Imparts superior stability and flow characteristics
  • Utilizes proteins that do not interact with mammalian antibodies
  • Supplied in HDPE plastic

Preservative - 0.09% sodium azide.

Applications: Western blot/EIA/Chemiluminescence.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty