Would you like to log in again?
Tube Storage Boxes, 50-Well Hinged
Supplier: Heathrow Scientific
120032 120033
77395-206CS 470.4 USD InStock
77395-206 77395-204
Tube Storage Boxes, 50-Well Hinged
Boxes Boxes for Tubes
Holds 1.5 and 2.0 ml microcentrifuge tubes and cryogenic vials.
  • Microtube storage box
  • Stylish and identifiable
  • Made of translucent, vibrantly colored polypropylene allows for visibility inside box and color coding identification
  • Tube capacity is indicated on the outside the box within a distinctive wave design
  • Quick visual identification with uniform wells and alphanumeric location identification along with a frosted labeling area
  • Robust
  • Easy open and secure lid fitted with either friction fit or snap clasp designs
  • Stackable, saving valuable space in the lab
The 50-well microtube storage box is ideal for sample or case study storage.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty