Would you like to log in again?
Tube Storage Box, 100-Well Hinged
Supplier: Heathrow Scientific
120043 120204 120203 120042 120202 120201
77395-214CS 349.92 USD InStock
77395-214 77395-222 77395-212 77395-220 77395-218 77395-216
Tube Storage Box, 100-Well Hinged
Boxes Boxes for Tubes
Holds 1.5 and 2.0 ml microcentrifuge tubes and cryogenic vials.
  • Microtube storage boxes
  • Hold 1.5 and 2.0 ml microcentrifuge tubes and cryogenic vials
  • Stylish and identifiable
  • Made of translucent, vibrantly colored polypropylene allows for visibility inside box and color coding identification
  • Tube capacity is indicated on the outside the box within a distinctive wave design
  • Quick visual identification with uniform wells and alphanumeric location identification along with a frosted labeling area
  • Robust
  • Easy open and secure lid fitted with either friction fit or snap clasp designs
  • Stackable, saving valuable space in the lab
The 100-well mcrotube storage box provides secure storage for microtubes and cryogenic vials.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty