Would you like to log in again?
Coated Wire Tube Rack
Supplier: Heathrow Scientific
120322
77395-234CS 712.8 USD InStock
77395-234
Coated Wire Tube Rack
Racks Tube Racks
Chemically resistant HDPE coated wire rack.
  • Holds all brands and types of conical, round, or flat bottom glass or plastic tubes
  • Square wells and three deck levels help keep tubes secure
Durable for transport or sample handling. It is also chemically resistant and suitable for conical, round, or flat bottom glass or plastic tubes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty