Would you like to log in again?
Reagent Reservoirs, 50 ml
Supplier: Heathrow Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
120645 120644 120643 120646
77395-266CS 656.1 USD InStock
77395-266 77395-264 77395-262 77395-260
Reagent Reservoirs, 50 ml
Reagent Reservoirs
Reagent Reservoirs by Heathrow Scientific offer a fresh and modern design with enhanced features to streamline pipetting procedures.
  • Superior durability with enhanced walls and crush-resistant 'turrets' at the corners
  • Easy handling with tabs to prevent stacked reservoirs from sticking together
  • Graduation markings on the top edge and inner walls allow measurements of liquid to be easily identified and reduce reagent waste
  • Pour-off spouts conveniently located on all four corners for easy spill-free pouring and reduced dripping
  • Divided reservoir design allows pipetting of two different reagents; maximize recovery if using only single channel and small amount of reagent
The 50 ml reagent reservoirs are available in sterile and non sterile formats. Sterile reagent reservoirs are manufactured and gamma irradiated in an ISO certified facility. They are certified Dnase-, Rnase-, and pyrogen-free.
Packaged individually in an easy open HDPE package. Traceability Certificate available upon request.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty