Would you like to log in again?
Low Temp Microscope Slide Box, 25-Place
Supplier: Avantor
120316V
77395-314CS 806.4 USD InStock
77395-314
Low Temp Microscope Slide Box, 25-Place
Microscope Slide Boxes
Store in ultra-low temperatures with confidence.
  • Holds standard slides (76×26 mm and 3×1")
  • Durable Construction
  • Made of polycarbonate to store in ultra-low temperatures
  • Uniform grooved slide slots separate slides preventing slide contact
  • Molded feet enable secure stacking for easy transport and storage
  • Durable Construction
  • Low temperature resistant numbered foam lining corresponds with identification sheet
  • Organize with 'Project' and 'Date' pre-molded labeling area
The low temperature microscope slide box protects your slides in ultra-low temperature ranges down to −80 °C.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty