Would you like to log in again?
Microscope Slide Boxes, 25-Place Foam
Supplier: Avantor
HS15989RV HS15989MV HS15989PV
77395-346CS 806.4 USD InStock
77395-346 77395-344 77395-342
Microscope Slide Boxes, 25-Place Foam
Microscope Slide Boxes
Easily access slides with friction-fit cover.
  • Organize and identify; durable and safe
  • Lining and slots are numbered to correspond to the slide inventory sheet
  • Organize by 'Project' and 'Date' with pre-molded labeling area
  • Available in multiple colors for project coding
  • ABS plastic boxes available with either cork or foam lining
  • Designed to securely store slides in grooved slots
  • Molded feet secure stacking for easy transport and storage
The premium microscope slide box is perfect for the storage or transportation of standard size slides (25×75 mm and 1×3").
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty