Would you like to log in again?
Microscope Slide Box, 100-Place
Supplier: Avantor
HS15994RV HS15994GV HS15994PV HS15994MV
77395-354CS 1267.2 USD InStock
77395-354 77395-352 77395-350 77395-348
Microscope Slide Box, 100-Place
Microscope Slide Boxes
Closes securely with a rust-resistant nickelplated clasp and pin hinges.
  • Organize and identify
  • Lining and slots are numbered to correspond to the slide inventory sheet
  • Organize by 'Project' and 'Date' with pre-molded labeling area
  • Available in multiple colors for project coding
  • Durable and safe
  • ABS plastic boxes available with either cork or foam lining
  • Designed to securely store slides in grooved slots
  • Molded feet secure stacking for easy transport and storage
The premium microscope slide box is perfect for the storage or transportation of standard size slides.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty