Would you like to log in again?
Bottle Rack, Coated Wire, 55 to 60 mm, 6-Place
Supplier: Heathrow Scientific
HS23224
77395-378EA 21.87 USD InStock
77395-378
Bottle Rack, Coated Wire, 55 to 60 mm, 6-Place
Racks Bottle Racks
Chemically resistant HDPE coated wire racks.
  • Holds all brands and types of conical, round or flat bottom, glass or plastic, tubes and/or bottles
  • Square wells and three deck levels help keep tubes secure
The HDPE coated wire rack is durable for transport or sample handling. It is also chemically resistant and suitable for conical, round or flat-bottom, or plastic tubes and bottles.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty