Would you like to log in again?
Fold and Snap Tube Racks, 17 mm, 60-Place
Supplier: Avantor
HS243077YV HS243077WV HS243077RV HS243077BV
77395-422CS 567 USD InStock
77395-422 77395-420 77395-418 77395-416
Fold and Snap Tube Racks, 17 mm, 60-Place
Racks Tube Racks
Economical durable rack.
  • Polypropylene racks ship flat in one piece
  • Easy-to-assemble, just fold and snap together
  • Stable design can be used in water baths
  • Open design allows samples to be incubated at the same temperature
  • Color code for easy identification
  • Easy to assemble just fold and snap together
The Fold and Snap Test tube rack is economical and durable.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty