Would you like to log in again?
Ice Pan Cool Container, 4 L, Blue
Supplier: Heathrow Scientific
HS28724B
77395-440CS 549.36 USD InStock
77395-440
Ice Pan Cool Container, 4 L, Blue
Ice Containers
Durable containers for chilling temperature-sensitive samples.
  • Suitable for low temperature use.; ideal for ice, ice slurries and CO₂ pellets
  • Made from polyurethane with metal inserts for stability
  • Close-fit lid keeps evaporation to a minimum and features a comfortable recessed knob
  • Tapered design for easy stacking
  • Precise pour spout and large handles
  • Lid nests under bucket for convenience and to save space
  • Large handles for safer transportation
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty