Would you like to log in again?
HyClone™ Defined Fetal Bovine Serum (FBS), New Zealand Origin
Supplier: HyClone products (Cytiva)
HyClone™
SH31194.02 SH31194.01 SH31194.03 SH31194.03HI SH31194.03IR30-45 SH31194.03 SH31194.01HI SH31194.01 SH31194.02 SH31194.03IR30-45 SH31194.01HI SH31194.02HI SH31194.02HI SH31194.03IR25-40 SH31194.03HI SH31194.03IR25-40
77395-478EA 359 USD InStock
CA77395-476 CA77395-486 77395-478 77395-486 CA77395-474 77395-476 CA77395-484 CA77395-472 77395-484 CA77395-482 77395-474 77395-482 77395-472 CA77395-480 CA77395-478 77395-480
HyClone™ Defined Fetal Bovine Serum (FBS), New Zealand Origin
Sera (Serum)
Highest quality FBS, HyClone™ AU-origin defined FBS is proprietary serial 40 nm-filtered, virus-tested to 9 CFR standard, and supplied with an extensive biochemical profile.
  • Well-defined: assay results for over 50 components, with additional testing available
  • NZ origin: sourced and processed in New Zealand with complete traceability to origin
  • Ultra-low endotoxin: ≤5 EU/ml endotoxin for reduced research and production risk
  • Sterile-filtered: Proprietary serial 40 nm filter process
γ-irradiated and heat-inactivated FBS options are available.
Certifications: Broad virus panel testing in accordance with 9 CFR 113.53.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty