Would you like to log in again?
GammaGuard CE® Sterile Coveralls with Attached Hood and Boots
Supplier: International Enviroguard
GammaGuard CE®
CE11019CIS-3XL CE11019CIS-2XL CE11019CIS-S CE11019CIS-L CE11019CIS-XL CE11019CIS-4XL CE11019CIS-M
77395-510CS 330.29 USD InStock
77395-510 77395-508 77395-516 77395-506 77395-514 77395-504 77395-512
GammaGuard CE® Sterile Coveralls with Attached Hood and Boots
Disposable Garments
Sterile coveralls are made from a comfortable fabric that protects against various liquids and particulates. Each coverall is individually packaged and sterilized to a 10⁻⁶ sterility assurance level (SAL).
  • Attached hood and boots eliminates the need for donning and storing separate hood and boot components
  • Attached thumb loops help prevent sleeves from riding up
  • Ankle ties provide customized fit for boots
  • Tunnelized elastic wrists, ankles, and back; serged seams
  • Sterile-Don™ packaging protects against contamination during donning
  • GammaGuard CE® helps keep wearer cooler compared to competitive fabrics
  • Meets high manufacturing standards; made in an ISO-certified facility
Coveralls are designed with tunnelized elastic that contains particle shed. Serged seams eliminate raw edges that can create and release additional particulate contamination. Work confidently with an attached hood and boots that provide more complete body protection. Elastic wrists and ankles help prevent particulates from entering or exiting the suit, while a storm flap over the front zipper closure adds an extra layer of protection. Convenient thumb loops are included to keep sleeves from riding up while ankle ties secure the attached boots.
Certifications: Certificate of sterility included in each case.
Packaging: Coveralls are  individually packaged, 25 per case.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty