Would you like to log in again?
Cryogenic Vial and Vaccine Vial Grippers
Supplier: Heathrow Scientific
120908 120909 120910
77395-610CS 3280 USD InStock
77395-610 77395-608 77395-606
Cryogenic Vial and Vaccine Vial Grippers
Forceps
The perfect grip for optimal precision.
  • Durable grippers are made of reinforced polypropylene, which is cold-tolerant and resistant to everyday wear
  • The designated squeeze target ensures maximum precision and grip with every use
  • Each style is available in a pack of 5 assorted colors for easy identification and organization
  • A combo pack with one of each style is also available
These two styles of hand-operated grippers are designed to pick up cryogenic and crimped 10R-size vaccine/serum vials, respectively.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty