Would you like to log in again?
Transport Tote XL for Samples
Supplier: Heathrow Scientific
120928 120927 120926
77395-616CS 336 USD InStock
77395-616 77395-614 77395-612
Transport Tote XL for Samples
Carriers Transport Carriers
High-capacity transport container for laboratory samples, instruments, and valuable products.
  • Sturdy and spacious, ideal for transporting a large volume of samples
  • Stainless steel, polypropylene, elastomeric compound, ABS construction
  • Transparent lid enables effortless identification of contents, reducing the risk of contamination
  • Equipped with articulated handles featuring silicone grips for comfortable and secure handling
A reliable solution for the safe and convenient transportation of laboratory samples, instruments, and valuable items. Crafted to meet the stringent demands of clinical and research laboratories, this oversized transport container ensures the protection and preservation of cargo.
A silicone seal ensures a tight enclosure, safeguarding samples from most environmental elements. Dual latches feature a security tag to guarantee a secure closure, providing tamper-evidenct security and ensuring the integrity and authenticity of samples. The bottom compartment is generously sized, allowing for carrying most tube racks, instruments or supplies according to specific requirements. Includes a MultiTube Rack Insert to accommodate various tube sizes. Additionally, two other insert options for blood tubes up to 10ml and general research tubes up to 15 ml are available separately.
Transparent design helps comply with OSHA 29 CFR 1910.1030 standards without having to re-label containers and enabling the user to visibly check sample integrity. This container meets UN3373 secondary packaging transport requirements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty