Would you like to log in again?
Transport Tote XL Tube Rack Inserts
Supplier: Heathrow Scientific
120960 120961 120962
77395-620CS 96 USD InStock
77395-620 77395-618 77395-622
Transport Tote XL Tube Rack Inserts
Racks Tube Racks
Inserts designed to fit the Transport tote XL sample transport container.
  • Thermoformed PETG construction
  • Tube Rack Inserts can be used separately
  • Lid removes completely for easy cleaning
  • Inserts are deep enough to be filled with ice
  • Non sterile
Rack inserts are available in small tube, large tube, and multi-tube formats, accommodating a wide variety of tube sizes and placement configurations.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty