Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ultra-Low Temperature Freezers, TSX Universal -86 °C ULT

Supplier: Thermo Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

TSX70086FARAKLN TSX40086FARAKLN TSX60086FARAKLB TSX50086LA TSX70086FARAK TSX70086LA TSX60086FARAKLN TSX40086FARAK TSX40086FARAKLB TSX50086FARAKCO TSX70086FARAKHID TSX40086FARAKCO TSX50086FARAKLN TSX60086FARAK TSX60086FA TSX60086FARAKHID TSX40086FA TSX50086FARAKHID TSX60086FARAKCO TSX50086FARAKLB TSX50086FARAK TSX40086FARAKHID TSX50086FA TSX70086FARAKLB TSX70086FARAKCO TSX60086LA TSX70086FA TSX40086LA
77400-784EA 29676.81 USD InStock
77400-784 77400-782 77400-788 77400-800 77400-786 77400-820 77400-780 77400-814 77400-834 77400-812 77400-818 77400-816 77400-796 77400-794 77400-772 77400-832 77400-810 77400-778 77400-830 77400-776 77400-798 77400-792 77400-804 77400-826 77400-802 77400-824 77400-808 77400-828
Ultra-Low Temperature Freezers, TSX Universal -86 °C ULT
Freezers Ultra Low Temperature Freezers
TSX Universal series ULT freezers are the pinnacle of reliability, sustainability, and innovation incorporating insights from over 80 years of experience to enhance every aspect of the system. Updates to the variable speed compressors has made the TSX Universal adaptable across all workflows. Available in four capacities, they are ENERGY STAR® certified with a 33% energy efficiency improvement, ACT labeled and produced in a Zero-Waste to Landfill certified facility.

  • Universal V-Drive technology delivers tighter peak variation, faster recovery times, decrease in energy consumption, improved pull down time
  • Improved user interface: Full 7" capacitive touch screen
  • Reduced rapid re-entry times: Improved pressure relief port enables re-entry in just 30 seconds helping ensure uninterrupted access to freezer
  • Expanded setpoint range: −40 to −86 °C
  • Universal voltage: Operates on 100 to 230 V, 50/60 Hz, streamlining procurement and qualification strategies from early research and development to global clinical trials
  • Simplified updates: Onboard software streamlines startup and integration, making it easier to access and install the latest updates for continuous access to new features
  • Industry-leading warranty: 12 years on compressor parts and five years unit

Whisper quiet noise under 44 dBA allows the TSX Universal ULT to be placed anywhere without causing sound pollution. Connectivity integration solutions helps monitor, share, and analyse data allowing streamlined processes and improved research outcomes.

By incorporating environmental sustainability principles into each design step, the environmental impact of these products is reduced early in the design process, helping to deliver on our commitment to achieve net-zero emissions by 2050 while helping customers advance their sustainability goals. Our strategy focuses on five key impact areas: Less hazardous, less waste, more energy efficient, responsibly packaged and extended life. All TSX Universal ULTs beat today’s ENERGY STAR criteria by wide margins and positioned to meet tightening requirements in the future.

Choice of four capacities: 400, 500, 600 and 700 (50.8 mm box)

Certifications: cULus, CE.

Ordering information: Accessories must be ordered separately. Field installed options and accessories must be installed by a qualified professional. Options include stainless steel interiors, five inner doors, racks, LN₂ and CO₂ back-up systems, status light, IQ/OQ documents and electronic locks.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

SERVICES

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty