Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Endeavor™ 5000 Light Duty Digital Orbital Shakers

Supplier: Ohaus
Endeavor™
30770910 30770909 30770911
77435-148EA 1000 USD InStock
77435-148 77435-146 77435-144
Endeavor™ 5000 Light Duty Digital Orbital Shakers
Shakers and Mixers Orbital Shakers
Digital orbital shakers you can trust for everyday shaking.

  • Budget-friendly: No need to buy a separate tray
  • Ready to attach a wide variety of accessories - customize for low to mid volume applications
  • Programmable: Save time and guarantee reproducible results
  • Simple on-board menu to set, store, and recall multi-step programs
  • Data logging and control: Remote function permits PC control and data transfer

Endeavor™ 5000 Light duty shakers are available in three models: 3 mm orbit, 20 mm orbit, or a dedicated microplate model. All models can be used in incubators and cold rooms. Stable, accurate, long-term shaking is ensured with the triple eccentric drive and maintenance-free brushless DC motor. Integrated overload and unbalanced detection to protect samples.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty