Would you like to log in again?
Rotary Microtome Knife Blocks
Supplier: Precisionary Instruments
RF600KB0TOP RF800KBBOTTOM RF1000KBMID RF800KBTOP RF600KBBOTTOM RF800KBMIDDLE RF600KBMIDDLE RF1000KBTOP RF800KBSET RF1000KBSET RF600KBSET RF1000KBBOTTOM
77435-346EA 865.1 USD InStock
77435-346 77435-302 77435-300 77435-332 77435-298 77435-352 77435-330 77435-350 77435-328 77435-326 77435-348 77435-304
Rotary Microtome Knife Blocks
Microtomes and Microtome Assemblies Microtome Blades and Handles
Rotary microtome knife blocks for use with models RF-1000, RF-800, and RF-600.
  • Knife block pieces are available individually or in sets
  • Knife block sets includes all three pieces
Rotary Microtome Knife Blocks are comprised of three pieces. The Bottom component allows for assembly onto microtome base. The middle component allows for angle adjustments. The top component features a cutting stage with blade guard and blade ejector.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty