Would you like to log in again?
Rotary Microtome Clamps
Supplier: Precisionary Instruments
RF600PARACLAMP RF800CASCLAMP RF600CASSCLAMP RF1000PARCLAMP RF1000CLAMP
77435-334EA 415.25 USD InStock
77435-334 77435-344 77435-320 77435-294 77435-306
Rotary Microtome Clamps
Microtomes and Microtome Assemblies Microtome Blades and Handles
An assortment of block clamps and cassette clamps for rotary microtome models RF-1000, RF-800, and RF-600.
  • Block and cassette clamps available for multiple rotary microtome models
Block clamps, or paraffin block clamps, fit the specimen head and can hold larger block samples for cutting. Casette holder clamps hold standard casettes for wax embedded tissue sectioning on the rotatry microtome.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty