Would you like to log in again?
Rotary Microtome Low Profile Blades
Supplier: Precisionary Instruments
RFBLADES
77435-360BX 249.63 USD InStock
77435-360
Rotary Microtome Low Profile Blades
Microtomes and Microtome Assemblies Microtome Blades and Handles
Rotary microtome low-profile cutting blades for tissue sectioning.
  • Box set contains 50 individual blades in a dispenser box
  • Blade angle: 35 degrees
These low profile blades are compatable with any rotary microtome model.
Packaging: Set includes one blade dispenser box with 50 blades.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty