Would you like to log in again?
Compresstome® Agarose tablets
Supplier: Precisionary Instruments
Compresstome®
VFAGTVM50CANAL VFAGTVM10CANAL VFAGT100CANAL VFAGTVM20CANAL
77435-388PK 324.51 USD InStock
77435-388 77435-394 77435-392 77435-390
Compresstome® Agarose tablets
Electrophoresis Gels Agarose and Acrylamide
Agarose tablets, each 0.5 mg, for easy use in specimen preparations for Compresstome® tissue samples.
  • Tablets are available in packs of 10, 20, 50, or 100
Agarose is used for fresh or fixed samples to section using a Compresstome® rotary microtome.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty