Would you like to log in again?
Compresstome® Legacy Model Blade Holder
Supplier: Precisionary Instruments
Compresstome®
VFBHVMLEGACYNC
77435-418EA 407.72 USD InStock
77435-418
Compresstome® Legacy Model Blade Holder
Microtomes and Microtome Assemblies Microtome Blades and Handles
Replacement blade holder for the Compresstome vibrating microtome.
  • Compatible with models VF-200, VF-200-0Z, VF-300, and VF-300-0Z
A replacement blade holder for legacy model Comprestomes. This device is compatible with Compresstome models VF-200, VF-200-0Z, VF-300, and VF-300-0Z. Includes one blade holder, one small Allen wrench, and one small screw.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty