Would you like to log in again?
Compresstome® Magnetic Blade Holder for VF-210-0Z and VF-510-0Z
Supplier: Precisionary Instruments
Compresstome®
VFBHVMMGNT
77435-420EA 574.14 USD InStock
77435-420
Compresstome® Magnetic Blade Holder for VF-210-0Z and VF-510-0Z
Microtomes and Microtome Assemblies Microtome Blades and Handles
This is a magnetic blade holder for easy stainless steel blade installation. This can also be used for ceramic blades.
  • Premium magnetic blade holder for the compresstome vibrating microtome
  • Each blade holder has built-in magnets to securely hold stainless steel blades without glue
  • Ceramic and tungsten blades can also be used with glue
Packaging: Includes one magnetic blade holder, one allen wrench, and one small screw.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty