Would you like to log in again?
Compresstome® Blades
Supplier: Precisionary Instruments
Compresstome®
VFBLVMCBCANAL VFBLVMSSBCANAL
77435-424EA 60.33 USD InStock
77435-424 77435-428
Compresstome® Blades
Microtomes and Microtome Assemblies Microtome Blades and Handles
Compresstome stainless steel or ceramic blades.
  • All blades are compatible with the compresstome models
Stainless steel blades are double-edged. They are packaged 5 double-edged blades per box (two boxes per order).
Ceramic blades for cutting tissue with compresstome slicers. Ceramic blades are durable and long-lasting, typically good for up to 3 to 4 weeks based on 1 sample per day. Sold as a single blade.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty