Would you like to log in again?
Compresstome® Gooseneck Lamp and Acessories
Supplier: Precisionary Instruments
Compresstome®
VFGLAMP VFMPLAMP
77435-464EA 199.7 USD InStock
77435-464 77435-472
Compresstome® Gooseneck Lamp and Acessories
Lights LED Lights, Lamps and Bulbs
The gooseneck lamp adds a spotlight when slicing specimens. The steel puck is used to attach the gooseneck lamp to the Compresstome® base.
  • LED spot illuminator lamp with flexible gooseneck
The Compresstome® lamp includes one LED spotlight on a flexible gooseneck that allows for easy adjustment. The steel puck allows for the magnet base of the Gooseneck Lamp to adhere when inserted on the Compresstome® vibrating microtome models.
Packaging: One set of screws/washers for mounting included. Steel puck with attached bolt includes one nut for securing bolt.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty