Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Air-Tite Premium 2-Part Lab Syringes, All Plastic, 50 ml

Supplier: Air-Tite
Air-Tite Premium
AML50 AMS50
77440-858BX 42.82 USD InStock
77440-858 77440-856
Air-Tite Premium 2-Part Lab Syringes, All Plastic, 50 ml
Syringes General-Purpose Syringes
These high-quality 50 ml 2-part syringes are made with polypropylene and polyethylene, without the rubber tip on the plunger or the silicone oil used most commonly in syringes.

  • Laboratory and veterinary use only
  • No rubber or silicone oil
  • Luer slip and lock accept common hypodermic fittings
  • Complies with ISO standards for syringes
  • Individually sterile packaged
  • Polypropylene barrel, polyethylene plunger

The largest in the Air-Tite line of 2-part, all-plastic syringes, these 50ml syringes are up to the task. Great for those applications which may be sensitive to the presence of silicone oil or rubber. Commonly used in everything from organic solvents to animal breeding to chromatography. Syringes successfully passed mouse embryo assay testing.

Certifications: Complies with ISO 7886.

Packaging: Individually sterile wrapped with 30 syringes per box, 12 boxes per case.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty