Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DuraClone Dry Unitized Antibody Panels For Flow Cytometry

Supplier: BECKMAN COULTER
DURAClone
C97741 C00162 B53318 B53328 C34369 C49589 C97741 C00162 B53328 B53309 C34369 B53318 C49589 B53346 B53346 B53309
77441-614EA 408.8 USD InStock
77441-614 CA77441-612 77441-616 CA77441-614 77441-620 77441-610 CA77441-620 CA77441-610 77441-612 CA77441-608 77441-606 CA77441-616 77441-618 77441-608 CA77441-606 CA77441-618
DuraClone Dry Unitized Antibody Panels For Flow Cytometry
Assays Cellular Assays
The dry DURAClone antibody panels are designed for clinical research studies. Every assay tube or well contains a layer with the dried-down antibody panel unitized for a single test.

  • Dry format, unitized for a single test, no antibody or stimulant pipetting
  • Flexible by optional addition of liquid reagents
  • Color compensation solution included (where required)
  • Expert-proven and thoroughly tested
  • Storage&shipping at room temperature
  • Simplified inventory management

Antibody panel development requires a high level of flow cytometry expertise and may consume critical laboratory resources. Take a leap forward and streamline panel implementation in your flow cytometry assay menu by using expert proven, dry DURAClone antibody combinations. The elimination of liquid antibody handling also efficiently reduces human error sources and reagent stability issues - the ideal solution for longitudinal and multi-center clinical research studies!

Certifications: Although not required for RUO status, this product is manufactured in accordance with Good Manufacturing Practices.

Delivery information: Supplied with screw cap and pouring ring.

Packaging: The bottles are packaged in shrinked-wrapped modules.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty