Would you like to log in again?
QuantiFluo™ Ammonia Assay Kit II
Supplier: BioAssay Systems
QuantiFluo™
D2NH3-100
77441-640EA 209 USD InStock
77441-640
QuantiFluo™ Ammonia Assay Kit II
Assays Cellular Assays
For quantitative determination of ammonia and ammonium in biological samples.
  • Fast and sensitive
  • Linear detection range of 0.005 to 2 mM ammonia
  • Convenient and high-throughput
  • A single component reagent
  • Homogeneous 'mix-incubate-measure' type assay
  • Can be readily automated on HTS liquid handling systems for processing thousands of samples per day
AMMONIA (NH₃) or its ion form ammonium (NH₄⁺) is an important source of nitrogen for living systems and is ubiquitously present in the nature. Simple, direct and automation-ready procedures for measuring NH₃ are very desirable. BioAssay Systems’ updated ammonia/ammonium assay is based on an improved o-phthalaldehyde method. This reagent reacts with ammonia/ammonium and forms a fluorescent product. The fluorescence intensity (λex/em = 415/475 nm) is proportional to the ammonia concentration in the sample.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty