Would you like to log in again?
Acetylcholinesterase Reagent
Supplier: BioAssay Systems
DACEN25ML
77441-644EA 89 USD InStock
77441-644
Acetylcholinesterase Reagent
Assays Enzyme Assays
Direct assays of acetylcholinesterase activity in blood, serum, plasma, and other biological samples.
  • Simple and convenient
  • This single reagent is ready to use, when reconstituted in 25 ml deionized water
  • High-throughput
  • Can be readily adapted on chemistry analyzers or high-throughput liquid handling system to test thousands of samples per day
ACETYLCHOLINESTERASE (EC 3.1.1.7, AChE), also known as RBC cholinesterase, is found primarily in the blood and neural synapses. Low serum cholinesterase activity may relate to exposure to insecticides or to one of a number of variant genotypes. AChE catalyzes the hydrolysis of the neurotransmitter acetylcholine into choline and acetic acid, a reaction necessary to allow a cholinergic neuron to return to its resting state after activation.
Cholinesterase levels of cells and plasma are used as a guide in establishing safety precautions relative to exposure and contact, as well as a guide in determining the need for workers to be removed from areas of contact with the organic phosphate insecticides. Simple, direct and automation-ready procedures for measuring AChE activity are very desirable. BioAssay Systems' acetylcholinesterase reagent is based on an improved Ellman method, in which thiocholine produced by the action of acetylcholinesterase forms a yellow color with 5,5’-dithiobis(2-nitrobenzoic acid). The intensity of the product color at 410 nm is proportionate to the enzyme activity in the sample.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty