Would you like to log in again?
QuantiFluo™ Asparaginase Assay Kit
Supplier: BioAssay Systems
QuantiFluo™
DASN-100
77441-652EA 359 USD InStock
77441-652
QuantiFluo™ Asparaginase Assay Kit
Assays Enzyme Assays
For quantitative determination of asparaginase activity in biological samples.
  • Safe and fast
  • Non-radioactive assay
  • Assay can be completed within 40 minutes
  • Sensitive and accurate
  • Linear detection range 1.1 to 300 U/L asparaginase in a 96-well plate assay
  • Convenient and high-throughput
  • Homogeneous 'mix-incubate-measure' type assay
  • Can be readily automated to assay thousands of samples per day
ASPARAGINASE (EC 3.5.1.1) is a key enzyme in the metabolism of asparagine. Its principal use is as treatment for acute lymphoblastic leukemia and lymphoblastic lymphoma where the asparagine is an essential amino acid. BioAssay Systems’ DASN-100 Kit provides a convenient fluorimetric method to measure asparaginase activity in biological samples. In this assay, o-phthalaldehyde reacts with liberated ammonia where the increase in fluorescence at λex/em = 415/475 nm is directly proportional to enzyme activity.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty