Would you like to log in again?
QuantiChrom™ Pyruvate Dehydrogenase Assay Kit
Supplier: BioAssay Systems
QuantiChrom™
DPDH-100
77441-660EA 389 USD InStock
77441-660
QuantiChrom™ Pyruvate Dehydrogenase Assay Kit
Assays Enzyme Assays
For assessing PDH activity in biological samples and screening and evaluation of PDH modulators.
  • High sensitivity and wide linear range
  • The detection limit is 0.87 U/L, with linearity up to at least 278 U/L PDH in a 96-well plate assay
  • Homogeneous and simple procedure
  • Simple 'mix-and-measure' procedure allows reliable quantitation of PDH activity within 20 minutes
  • Robust and amenable to HTS
  • All reagents are compatible with high-throughput liquid handling instruments
PYRUVATE DEHYDROGENASE (PDH) (E.C. 1.2.4.1) is an oxidoreductase which catalyzes the decarboxylation of pyruvate to form acetylated dihydrolipoamide in a thiamine pyrophosphate (TPP)-dependent reaction. PDH is the E1 component of the PDH complex that ultimately produces acetyl-CoA. The PDH complex connects glycolysis to the citric acid cycle. This non-radioactive colorimetric PDH assay is based on the reduction of the tetrazolium salt MTT in a Phenazine Methosulfate (PMS) coupled reaction to a reduced form of MTT which exhibits an absorption maximum at 565 nm. The intensity of the purple color formed is directly proportional to the PDH activity.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty