Would you like to log in again?
QuantiChrom™ Pectinase Assay Kit
Supplier: BioAssay Systems
QuantiChrom™
DPEC-100
77441-662EA 309 USD InStock
77441-662
QuantiChrom™ Pectinase Assay Kit
Assays Enzyme Assays
For quantitative determination of pectinase activity determination in biological samples.
  • Linear detection range from 9.2 to 100 U/L pectinase activity in a 96-well plate assay
  • Simple and convenient 40 - minute 'add-mix-measure' type assay
  • High-throughput format and compatible with laboratory liquid handling systems
  • No heating is required
  • The kit does not use toxic materials
  • Refer to the previously established DNS method
Pectinase (EC 3.2.1.15) is an enzyme that catalyzes the hydrolysis of pectin, a heteropolysaccharide found primarily in the middle lamella and cell wall of terrestrial plants. During fruit ripening, enzymes, including pectinase break down the middle lamella, separating cells and softening the fruit. Pectinase enzymes are used in the food industry to liquefy fruit pulp and clarify fruit juice and wine.
BioAssay systems DPEC-100 kit provides a rapid and convenient assay for pectinase enzyme activity, in which pectin the substrate forms a turbid complex with the detection reagent. The turbidity, measured at OD600 nm, is proportional to the amount of unhydrolyzed pectin, thus inversely proportional to the pectinase activity.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty