Would you like to log in again?
QuantiChrom™ Plasmin Assay Kit
Supplier: BioAssay Systems
QuantiChrom™
DPLM-100
77441-664EA 429 USD InStock
77441-664
QuantiChrom™ Plasmin Assay Kit
Assays Protein Assays
For quantitative determination of plasmin enzyme activity in biological samples.
  • Safe and sensitive
  • Non-radioactive assay
  • Use as little as 10 μl samples
  • Linear detection range in 96-well plate: 0.35 to 56.5 U/L activity
  • Fast and convenient
  • The procedure involves addition of a single working reagent and incubation for 30 min
  • High-throughput
  • Homogeneous 'mix-incubate-measure' type assay
  • Can be readily automated to assay thousands of samples per day
PLASMIN (3.4.21.7) is a serine protease that is a key enzyme in the process of fibrinolysis. Plasmin is released into circulation as its inactive form, plasminogen, which can be activated by enzymes such as urokinase. The plasmin/plasminogen system has been studied for its role in inflammation, degradation of the extracellular matrix, and wound healing. BioAssay Systems’ DPLM-100 kit provides a convenient colorimetric method to measure plasmin activity. In this assay, plasmin hydrolyzes a synthetic substrate to release p-nitroanilide (pNA), which absorbs at 405 nm. The increase in absorbance at 405 nm is directly proportional to the enzyme activity.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty