Would you like to log in again?
QuantiChrom™ Pyrophosphatase Inhibitor Assay Kit
Supplier: BioAssay Systems
QuantiChrom™
DPPI-100
77441-666EA 359 USD InStock
77441-666
QuantiChrom™ Pyrophosphatase Inhibitor Assay Kit
Assays Enzyme Assays
For high-throughput inhibitor screening and evaluation of pyrophosphatase modulators.
  • Safe and non-radioactive assay
  • Fast and convenient
  • The procedure involves incubation of enzyme and inhibitor, addition of a single working reagent and incubation for 30 min
  • Room temperature assay
  • No 37 °C incubator is needed
  • High-throughput
  • Homogenous 'mix-incubate-measure' type assay
  • Can be readily automated on HTS liquid handling systems
  • Robust assay with a Z' factor of 0.59
  • Can be used in 96-well, 384-well, and potentially higher density screening assays
Inorganic pyrophosphatase (E.C.3.6.1.1) catalyzes the hydrolysis of phosphoester bonds of inorganic pyrophosphate [P2O74-], thereby releasing two orthophosphate molecules. Family I PPases are essential enzymes found in all kingdoms of life and are responsible for maintaining the correct pyrophosphate equilibrium necessary to carry out nucleic acid and protein synthesis, and facilitate fatty acid beta oxidation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty