Would you like to log in again?
QuantiFluo™ Trypsin Inhibitor Assay Kit
Supplier: BioAssay Systems
QuantiFluo™
DTRI-100
77441-670EA 439 USD InStock
77441-670
QuantiFluo™ Trypsin Inhibitor Assay Kit
Assays Enzyme Assays
For evaluation of drugs and screening of potential inhibitors to trypsin proteases.
  • Safe and non-radioactive assay
  • Homogeneous and convenient
  • Homogenous 'mix-incubate-measure' type assay
  • No wash and reagent transfer steps are involved
  • Robust and high-throughput
  • Can be readily automated to assay thousands of samples per day
  • Robust assay with a Z factor of >0.7
Trypsin (EC 3.4.21.4) is a digestive, serine protease that hydrolyzes dietary proteins in many eukaryotic and prokaryotic organisms. Trypsin predominantly cleaves peptide chains at the carboxyl side of lysine and arginine amino acids, but not before proline. BioAssay systems QuantiFluo™ trypsin inhibitor assay kit uses a fluorescein isothiocyanate (FITC)-labeled synthetic substrate.
The fluorescein label is highly quenched. Upon digestion by trypsin present in the sample, the substrate is cleaved into smaller peptides, which abolishes the quenching of the fluorescence label. The fluorescence or fluorescence polarization (FP) of the FITC-labeled fragments is measured at λex/em = 485/530 nm. Inhibition is determined by the decrease in fluorescence.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty