Would you like to log in again?
QuantiFluo™ Urokinase Inhibitor Screening Assay Kit
Supplier: BioAssay Systems
QuantiFluo™
DUKI-100
77441-672EA 399 USD InStock
77441-672
QuantiFluo™ Urokinase Inhibitor Screening Assay Kit
Assays Enzyme Assays
For evaluation of drugs and screening potential inhibitors of urokinase.
  • Safe
  • Non-radioactive assay
  • Fast
  • Assay is completed within a 30 minutes reaction time
  • Homogeneous 'mix-incubate-measure' type assay
  • Can be readily automated to assay thousands of samples per day
UROKINASE PLASMINOGEN ACTIVATOR (urokinase, uPA) is a key serine protease involved in the degradation of the extracellular matrix that catalyzes the conversion of plasminogen to active plasmin. It acts as a thrombolytic agent to break up blood clots and when over-expressed, has been reported to influence the growth of certain malignant tumors (breast, prostate, etc.). BioAssay Systems’ DUKI-100 Kit provides a convenient fluorimetric means to screen for potential urokinase inhibitors. In this assay, the fluorimetric substrate reacts with urokinase and an inhibitor will decrease the fluorescence at λex/em = 380/450 nm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty