Would you like to log in again?
QuantiFluo™ Urokinase Assay Kit
Supplier: BioAssay Systems
QuantiFluo™
DUKN-100
77441-674EA 399 USD InStock
77441-674
QuantiFluo™ Urokinase Assay Kit
Assays Enzyme Assays
For quantitative determination of urokinase activity determination in biological samples.
  • Safe and non-radioactive assay
  • Fast
  • Assay is completed within a 15 minute reaction time
  • Homogeneous 'mix-incubate-measure' type assay
  • Can be readily automated to assay thousands of samples per day
Urokinase plasminogen activator (urokinase, uPA) is a key serine protease involved in the degradation of the extracellular matrix that catalyzes the conversion of plasminogen to active plasmin. It acts as a thrombolytic agent to break up blood clots and when over-expressed, has been reported to influence the growth of certain malignant tumors (breast, prostate, etc).
BioAssay systems DUKN-100 kit provides a convenient fluorimetric method to measure urokinase activity in biological samples. In this assay, the fluorimetric substrate reacts with urokinase so that the increase in fluorescence at λex/em = 380/450 nm is directly proportional to enzyme activity.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty