Would you like to log in again?
EnzyChrom™ Creatinine Assay Kit
Supplier: BioAssay Systems
EnzyChrom™
E2CT-100
77441-676EA 409 USD InStock
77441-676
EnzyChrom™ Creatinine Assay Kit
Assays Enzyme Assays
For quantitative determination of creatinine and evaluation of drug effects on its metabolism.
  • Fast and sensitive
  • Linear detection range: 4.8 to 500 µM or 0.054 to 5.7 mg/dl (colorimetric assay) and 0.25 to 100 µM or 0.0028 to 1.14 mg/dl (fluorimetric assay) for a 60 min reaction
  • It is 3- and 53-fold more sensitive than the traditional Jaffe method (e.g. DICT-500), especially useful for small samples or where high sensitivity is required
  • Convenient
  • The procedure involves adding a single working reagent and reading after 60 minutes
  • Room temperature assay
  • No 37 °C heater is needed
  • High-throughput
  • Homogeneous 'mix-incubate-measure' type assay
  • Can be readily automated to process thousands of samples per day
CREATININE is synthesized in the body from creatine, which is produced during muscle contractions from creatine phosphate. In the blood, creatinine is removed by filtration through the glomeruli of the kidney and is secreted into urine. In healthy individuals, creatinine secretion is independent of diet and is fairly constant. The creatinine clearance test has become one of the most sensitive tests for measuring glomerular filtration rate.
In kidney disease, creatinine levels in the blood are elevated, whereas the creatinine clearance rate and hence the urine levels are diminished. Creatinine testing is most widely used to assess kidney function. BioAssay Systems’ enzyme-based creatinine assay uses a reaction sequence that excludes endogenous ammonia to convert a dye into a colored and fluorescent form. The absorbance at 570 nm or fluorescence intensity at λex/em = 530/585 nm is directly proportional to the creatinine concentration in the sample.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty